Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191625.1 | NP_001178554.1 | 5 - 6 | 834 | 131 | |
XM_006229375.2 | XP_006229437.1 | 6 - 7 | 1183 | 131 | ||
XM_008759522.1 | XP_008757744.1 | 6 - 7 | 1184 | 131 | ||
XM_008759523.1 | XP_008757745.1 | 6 - 7 | 1512 | 131 | ||
XM_008759524.1 | XP_008757746.1 | 7 - 8 | 1257 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |