Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106340.1 | NP_001099810.1 | 7 - 8 | 1471 | 145 | |
GenBank mRNA | BC161909.1 | - | 7 - 8 | 1444 | 8367841 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |