Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183053.1 | NP_898876.1 | 6 - 7 | 1031 | 68 | |
GenBank mRNA | AY129073.1 | - | 6 - 7 | 1031 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |