Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057207.3 | NP_476555.1 | 7 - 8 | 1558 | 65 | |
XM_008759492.1 | XP_008757714.1 | 7 - 8 | 2277 | 65 | ||
XM_008759493.1 | XP_008757715.1 | 7 - 8 | 2655 | 65 | ||
XM_008759494.1 | XP_008757716.1 | 7 - 8 | 3177 | 65 | ||
XM_008759495.1 | XP_008757717.1 | 7 - 8 | 2273 | 65 | ||
GenBank mRNA | L10362.1 | - | 7 - 8 | 1558 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |