Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107525.2 | NP_001100995.1 | 3 - 4 | 246 | 149 | |
XM_006229430.2 | XP_006229492.1 | 2 - 3 | 592 | 149 | ||
GenBank mRNA | BC158658.1 | - | 3 - 4 | 246 | 8369176 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |