Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107534.1 | NP_001101004.1 | 5 - 6 | 449 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |