Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253318.2 | XP_006253380.1 | 9 - 10 | 1085 | 56 | |
XM_008771354.1 | XP_008769576.1 | 11 - 12 | 2081 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |