Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276712.1 | NP_001263641.1 | 1 - 2 | 539 | 94 | |
GenBank mRNA | AF167308.1 | - | 1 - 2 | 135 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |