Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107321.1 | NP_001100791.1 | 4 - 5 | 1122 | 67 | |
XM_008771355.1 | XP_008769577.1 | 4 - 5 | 4286 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |