Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107320.1 | NP_001100790.1 | 2 - 3 | 454 | 88 | |
XM_008771329.1 | XP_008769551.1 | 2 - 3 | 260 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |