Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230922.2 | XP_006230984.1 | 4 - 5 | 495 | 81 | |
XM_006230923.2 | XP_006230985.1 | 4 - 5 | 755 | 81 | ||
XM_006230924.2 | XP_006230986.1 | 4 - 5 | 552 | 81 | ||
XM_006230925.2 | XP_006230987.1 | 4 - 5 | 519 | 81 | ||
XM_008760190.1 | XP_008758412.1 | 4 - 5 | 398 | 81 | ||
XM_574595.6 | XP_574595.3 | 4 - 5 | 1076 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |