Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022407.3 | NP_071852.2 | 4 - 5 | 474 | 77 | |
GenBank mRNA | AF001896.1 | - | 4 - 5 | 474 | 77 | |
AF001897.1 | - | 4 - 5 | 474 | 77 | ||
AF001898.1 | - | 4 - 5 | 474 | 77 | ||
BC061526.1 | - | 4 - 5 | 493 | 6921926 | 77 | |
L42009.1 | - | 4 - 5 | 470 | 77 | ||
U79118.1 | - | 4 - 5 | 473 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |