Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134428.1 | NP_001127900.1 | 1 - 2 | 100 | 88 | |
XM_006230757.1 | XP_006230819.1 | 1 - 2 | 152 | 88 | ||
GenBank mRNA | BG669308.1 | - | 1 - 2 | 100 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |