Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108925.2 | NP_001102395.1 | 6 - 7 | 418 | 99 | |
GenBank mRNA | BC158732.1 | - | 6 - 7 | 418 | 9092746 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |