Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106375.2 | NP_001099845.2 | 7 - 8 | 900 | 138 | |
GenBank mRNA | BC169061.1 | - | 7 - 8 | 900 | 5621354 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |