Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271229.1 | NP_001258158.1 | 1 - 2 | 253 | 150 | |
GenBank mRNA | BC168162.1 | - | 1 - 2 | 253 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |