Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191083.1 | NP_001178012.1 | 10 - 11 | 702 | 152 | |
GenBank mRNA | FQ230255.1 | - | 7 - 8 | 459 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |