Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115048.1 | NP_001108520.1 | 6 - 7 | 1075 | 96 | |
XM_006231296.2 | XP_006231358.1 | 6 - 7 | 1122 | 96 | ||
XM_006231297.2 | XP_006231359.1 | 6 - 7 | 887 | 96 | ||
XM_006231299.2 | XP_006231361.1 | 6 - 7 | 1259 | 96 | ||
XM_008760346.1 | XP_008758568.1 | 5 - 6 | 844 | 96 | ||
XM_008760347.1 | XP_008758569.1 | 4 - 5 | 1100 | 96 | ||
XM_008760349.1 | XP_008758571.1 | 5 - 6 | 875 | 96 | ||
GenBank mRNA | BC158624.1 | - | 6 - 7 | 1075 | 7454746 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |