Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138887.1 | NP_620242.1 | 2 - 3 | 324 | 91 | |
GenBank mRNA | D29960.1 | - | 2 - 3 | 324 | 91 | |
FQ215336.1 | - | 2 - 3 | 353 | 91 | ||
FQ217608.1 | - | 2 - 3 | 353 | 91 | ||
FQ224974.1 | - | 2 - 3 | 353 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |