Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759423.1 | XP_008757645.1 | 2 - 3 | 605 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |