Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106234.1 | NP_001099704.1 | 5 - 6 | 645 | 83 | |
GenBank mRNA | BC158848.1 | - | 5 - 6 | 605 | 7444454 | 83 |
FQ209618.1 | - | 5 - 6 | 616 | 83 | ||
FQ221207.1 | - | 1 - 2 | 57 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |