Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004075.1 | NP_001004075.1 | 5 - 6 | 1055 | 99 | |
XM_006249360.2 | XP_006249422.1 | 5 - 6 | 1588 | 99 | ||
XM_006249361.2 | XP_006249423.1 | 6 - 7 | 1265 | 99 | ||
XM_008769214.1 | XP_008767436.1 | 5 - 6 | 973 | 99 | ||
GenBank mRNA | AY157968.1 | - | 4 - 5 | 960 | 99 | |
BC079044.1 | - | 5 - 6 | 1055 | 7115027 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |