Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107137.1 | NP_001100607.1 | 10 - 11 | 1388 | 55 | |
GenBank mRNA | BC168924.1 | - | 10 - 11 | 1370 | 9029270 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |