Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107137.1 | NP_001100607.1 | 3 - 4 | 390 | 83 | |
GenBank mRNA | BC168924.1 | - | 3 - 4 | 372 | 9029270 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |