Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107137.1 | NP_001100607.1 | 8 - 9 | 1200 | 82 | |
GenBank mRNA | BC168924.1 | - | 8 - 9 | 1182 | 9029270 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |