Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127570.1 | NP_001121042.1 | 1 - 2 | 261 | 112 | |
GenBank mRNA | BC161996.1 | - | 1 - 2 | 261 | 9034684 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |