Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012897.2 | NP_037029.1 | 5 - 6 | 449 | 74 | |
XM_008769317.1 | XP_008767539.1 | 4 - 5 | 340 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |