Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105920.1 | NP_001099390.1 | 1 - 2 | 135 | 94 | |
GenBank mRNA | BC166568.1 | - | 1 - 2 | 118 | 7318881 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |