Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108332.1 | NP_001101802.1 | 9 - 10 | 1271 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |