Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC158866.1 | - | 7 - 8 | 1021 | 5620924 | 99 |
FQ212189.1 | - | 7 - 8 | 1034 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |