Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105933.1 | NP_001099403.1 | 2 - 3 | 214 | 96 | |
GenBank mRNA | BC166554.1 | - | 2 - 3 | 173 | 7311069 | 96 |
FQ219862.1 | - | 2 - 3 | 188 | 96 | ||
FQ221109.1 | - | 2 - 3 | 174 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |