Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019237.1 | NP_062110.1 | 8 - 9 | 1306 | 96 | |
GenBank mRNA | AB008534.1 | - | 8 - 9 | 1293 | 96 | |
AF016503.1 | - | 8 - 9 | 1300 | 96 | ||
BC086572.1 | - | 9 - 10 | 1362 | 7304047 | 96 | |
U94710.1 | - | 8 - 9 | 1306 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |