Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107145.1 | NP_001100615.1 | 7 - 8 | 1210 | 73 | |
XM_008769233.1 | XP_008767455.1 | 8 - 9 | 1395 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |