Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 10 - 11 | 1704 | 80 | |
XM_006251928.2 | XP_006251990.1 | 9 - 10 | 2556 | 80 | ||
XM_008770661.1 | XP_008768883.1 | 8 - 9 | 1517 | 80 | ||
XM_008770662.1 | XP_008768884.1 | 9 - 10 | 1888 | 80 | ||
GenBank mRNA | AF036537.2 | - | 10 - 11 | 1704 | 80 | |
BC158580.1 | - | 8 - 9 | 1271 | 7378607 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |