Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006962.1 | NP_001006963.1 | 6 - 7 | 1220 | 87 | |
XM_006251940.2 | XP_006252002.1 | 6 - 7 | 2208 | 87 | ||
XM_006251942.2 | XP_006252004.1 | 6 - 7 | 2208 | 87 | ||
XM_006251943.2 | XP_006252005.1 | 6 - 7 | 2209 | 87 | ||
XM_006251944.2 | XP_006252006.1 | 6 - 7 | 2208 | 87 | ||
GenBank mRNA | BC083766.1 | - | 6 - 7 | 1220 | 7193196 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |