Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169139.1 | NP_001162610.1 | 5 - 6 | 1305 | 151 | |
GenBank mRNA | CK596035.1 | - | 2 - 3 | 543 | 151 | |
EV778235.1 | - | 2 - 3 | 264 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |