Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106088.2 | NP_001099558.1 | 7 - 8 | 542 | 96 | |
XM_008771323.1 | XP_008769545.1 | 7 - 8 | 795 | 96 | ||
XM_008771324.1 | XP_008769546.1 | 7 - 8 | 737 | 96 | ||
GenBank mRNA | BC168668.1 | - | 7 - 8 | 542 | 7934033 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |