Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253451.2 | XP_006253513.1 | 5 - 6 | 857 | 89 | |
XM_008771397.1 | XP_008769619.1 | 6 - 7 | 860 | 89 | ||
XM_008771398.1 | XP_008769620.1 | 6 - 7 | 939 | 89 | ||
XM_008771399.1 | XP_008769621.1 | 6 - 7 | 767 | 89 | ||
XM_008771400.1 | XP_008769622.1 | 5 - 6 | 893 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |