Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025718.1 | NP_001020889.1 | 6 - 7 | 814 | 85 | |
XM_006253451.2 | XP_006253513.1 | 7 - 8 | 1079 | 85 | ||
XM_008771397.1 | XP_008769619.1 | 8 - 9 | 1082 | 85 | ||
XM_008771398.1 | XP_008769620.1 | 8 - 9 | 1161 | 85 | ||
XM_008771399.1 | XP_008769621.1 | 8 - 9 | 989 | 85 | ||
XM_008771400.1 | XP_008769622.1 | 7 - 8 | 1115 | 85 | ||
GenBank mRNA | BC098789.1 | - | 6 - 7 | 814 | 7375514 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |