Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134744.1 | NP_001128216.1 | 4 - 5 | 486 | 103 | |
XM_006253381.2 | XP_006253443.1 | 4 - 5 | 758 | 103 | ||
GenBank mRNA | BC168222.1 | - | 4 - 5 | 486 | 7323695 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |