Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138532.1 | NP_612541.1 | 5 - 6 | 564 | 59 | |
XM_008769600.1 | XP_008767822.1 | 3 - 4 | 527 | 59 | ||
XM_008769601.1 | XP_008767823.1 | 3 - 4 | 529 | 59 | ||
GenBank mRNA | AF202049.1 | - | 5 - 6 | 564 | 59 | |
BC060314.1 | - | 5 - 6 | 585 | 6918381 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |