Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145683.1 | NP_663716.1 | 9 - 10 | 1090 | 114 | |
XM_006249820.2 | XP_006249882.1 | 9 - 10 | 1565 | 114 | ||
XM_006249821.2 | XP_006249883.1 | 9 - 10 | 1220 | 114 | ||
GenBank mRNA | U28356.1 | - | 9 - 10 | 1090 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Rattus norvegicus 1700 Probe spans exons |