Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012516.2 | NP_036648.2 | 8 - 9 | 1158 | 116 | |
GenBank mRNA | BC088165.1 | - | 8 - 9 | 1158 | 7324823 | 116 |
Z50051.1 | - | 6 - 7 | 821 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |