Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005871.1 | NP_001005871.1 | 1 - 2 | 363 | 101 | |
XM_008769446.1 | XP_008767668.1 | 2 - 3 | 746 | 101 | ||
XM_008769447.1 | XP_008767669.1 | 2 - 3 | 1383 | 101 | ||
XM_008769448.1 | XP_008767670.1 | 2 - 3 | 746 | 101 | ||
XM_008769449.1 | XP_008767671.1 | 2 - 3 | 747 | 101 | ||
XM_008769450.1 | XP_008767672.1 | 2 - 3 | 747 | 101 | ||
GenBank mRNA | U15408.1 | - | 1 - 2 | 363 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Rattus norvegicus Probe spans exons |