Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005871.1 | NP_001005871.1 | 15 - 16 | 2769 | 55 | |
XM_008769446.1 | XP_008767668.1 | 16 - 17 | 3152 | 55 | ||
XM_008769447.1 | XP_008767669.1 | 16 - 17 | 3789 | 55 | ||
XM_008769448.1 | XP_008767670.1 | 15 - 16 | 3116 | 55 | ||
XM_008769449.1 | XP_008767671.1 | 16 - 17 | 3153 | 55 | ||
XM_008769450.1 | XP_008767672.1 | 15 - 16 | 3117 | 55 | ||
GenBank mRNA | U15408.1 | - | 15 - 16 | 2769 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Rattus norvegicus Probe spans exons |