Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047902.1 | NP_001041367.1 | 14 - 15 | 2203 | 63 | |
XM_008769482.1 | XP_008767704.1 | 12 - 13 | 1980 | 63 | ||
XM_008769483.1 | XP_008767705.1 | 12 - 13 | 1977 | 63 | ||
XM_008769484.1 | XP_008767706.1 | 12 - 13 | 1953 | 63 | ||
GenBank mRNA | BC111404.1 | - | 7 - 8 | 1216 | 63 | |
DQ480752.1 | - | 14 - 15 | 2203 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |