Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025126.1 | NP_001020297.1 | 9 - 10 | 1131 | 72 | |
XM_006250113.2 | XP_006250175.1 | 16 - 17 | 2330 | 72 | ||
XM_006250114.2 | XP_006250176.1 | 16 - 17 | 2318 | 72 | ||
XM_006250115.2 | XP_006250177.1 | 15 - 16 | 2270 | 72 | ||
XM_006250116.2 | XP_006250178.1 | 15 - 16 | 2258 | 72 | ||
GenBank mRNA | BC091284.1 | - | 9 - 10 | 1131 | 7304516 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |