Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012676.1 | NP_036808.1 | 8 - 9 | 421 | 102 | |
XM_006249822.1 | XP_006249884.1 | 10 - 11 | 530 | 102 | ||
XM_006249823.1 | XP_006249885.1 | 10 - 11 | 531 | 102 | ||
XM_006249825.2 | XP_006249887.1 | 10 - 11 | 544 | 102 | ||
XM_006249828.1 | XP_006249890.1 | 9 - 10 | 518 | 102 | ||
XM_006249829.1 | XP_006249891.1 | 9 - 10 | 519 | 102 | ||
XM_006249833.2 | XP_006249895.1 | 9 - 10 | 526 | 102 | ||
XM_006249835.1 | XP_006249897.1 | 8 - 9 | 488 | 102 | ||
XM_006249836.1 | XP_006249898.1 | 8 - 9 | 489 | 102 | ||
XM_006249838.1 | XP_006249900.1 | 8 - 9 | 489 | 102 | ||
XM_006249840.2 | XP_006249902.1 | 11 - 12 | 560 | 102 | ||
XM_006249842.2 | XP_006249904.1 | 9 - 10 | 517 | 102 | ||
XM_008769535.1 | XP_008767757.1 | 10 - 11 | 531 | 102 | ||
XM_008769536.1 | XP_008767758.1 | 10 - 11 | 531 | 102 | ||
XM_008769537.1 | XP_008767759.1 | 9 - 10 | 519 | 102 | ||
XM_008769538.1 | XP_008767760.1 | 8 - 9 | 489 | 102 | ||
GenBank mRNA | BC161855.1 | - | 9 - 10 | 526 | 9035753 | 102 |
EU295527.1 | - | 7 - 8 | 391 | 102 | ||
M26051.1 | - | 8 - 9 | 421 | 102 | ||
M26052.1 | - | 8 - 9 | 441 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |