Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153730.2 | NP_714952.1 | 2 - 3 | 358 | 89 | |
XM_006249843.2 | XP_006249905.1 | 3 - 4 | 507 | 89 | ||
XM_006249844.2 | XP_006249906.1 | 3 - 4 | 781 | 89 | ||
XM_006249845.1 | XP_006249907.1 | 3 - 4 | 487 | 89 | ||
XM_006249846.2 | XP_006249908.1 | 4 - 5 | 573 | 89 | ||
XM_008769539.1 | XP_008767761.1 | 3 - 4 | 546 | 89 | ||
XM_008769540.1 | XP_008767762.1 | 4 - 5 | 401 | 89 | ||
XM_008769541.1 | XP_008767763.1 | 3 - 4 | 291 | 89 | ||
GenBank mRNA | AJ133126.1 | - | 2 - 3 | 400 | 89 | |
BC086983.1 | - | 3 - 4 | 488 | 7133597 | 89 | |
FQ210179.1 | - | 4 - 5 | 557 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |