Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133590.1 | NP_598274.1 | 3 - 4 | 726 | 64 | |
XM_006249744.2 | XP_006249806.1 | 4 - 5 | 703 | 64 | ||
XM_006249745.2 | XP_006249807.1 | 4 - 5 | 560 | 64 | ||
XM_008769422.1 | XP_008767644.1 | 2 - 3 | 256 | 64 | ||
GenBank mRNA | BC081732.1 | - | 4 - 5 | 682 | 7109012 | 64 |
FQ233577.1 | - | 3 - 4 | 492 | 64 | ||
FQ234855.1 | - | 3 - 4 | 492 | 64 | ||
X96663.1 | - | 3 - 4 | 726 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |